3-Ethynyl-2-methoxypyridine 97%

Lieferant: Apollo Scientific

Synonyme: 3-Ethynylpyridin-2-yl methyl ether, (2-Methoxypyridin-3-yl)acetylene

OR52205-250MG
APOSOR52205-250MGEA 128 EUR
APOSOR52205-250MG
3-Ethynyl-2-methoxypyridine 97%
3-Ethynyl-2-methoxypyridine
Formel: C₈H₇NO
Molecular Weight: 133,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189765
CAS-Nummer: 1196145-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR