Methyl 5-ethynylpyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 5-ethynylpicolinate, 5-Ethynyl-2-(methoxycarbonyl)pyridine

OR52206-100MG
APOSOR52206-100MGEA 21.9 EUR
APOSOR52206-100MG
Methyl 5-ethynylpyridine-2-carboxylate
Methyl 5-ethynylpyridine-2-carboxylate
Formel: C₉H₇NO₂
Molecular Weight: 161.157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189756
CAS-Nummer: 17880-61-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR