3-Chloro-4-morpholin-4-ylphenylamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-4-(morpholin-4-yl)aniline
OR5221-1G
APOSOR5221-1GEA
94.5
EUR
APOSOR5221-1G
3-Chloro-4-morpholin-4-ylphenylamine
3-Chloro-4-morpholin-4-ylphenylamine
Formel:
C₁₀H₁₃ClN₂O Molecular Weight: 212,68 g/mol Schmelzpunkt: 103…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047411 CAS-Nummer: 55048-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...