(1-Methyl-1H-[1,2,4]triazol-3-yl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
OR52221-5G
APOSOR52221-5GEA
150
EUR
APOSOR52221-5G
(1-Methyl-1H-[1,2,4]triazol-3-yl)methanol
(1-Methyl-1H-[1,2,4]triazol-3-yl)methanol
Formel:
C₄H₇N₃O Schmelzpunkt: 72…75 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD16710296 CAS-Nummer: 135242-93-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...