3-Amino-5-bromo-2-methoxybenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-3-carboxy-2-methoxyaniline

OR52230-1G OR52230-250MG OR52230-5G
APOSOR52230-1GEA 986 EUR
APOSOR52230-1G APOSOR52230-250MG APOSOR52230-5G
3-Amino-5-bromo-2-methoxybenzoic acid
3-Amino-5-bromo-2-methoxybenzoic acid
Formel: C₈H₈BrNO₃
Molecular Weight: 246.058 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28384162
CAS-Nummer: 60541-85-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR