2-Methyl-1,6-naphthyridine-3-carboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR5224-250MG
APOSOR5224-250MGEA 29 EUR
APOSOR5224-250MG
2-Methyl-1,6-naphthyridine-3-carboxylic acid 95+%
2-Methyl-1,6-naphthyridine-3-carboxylic acid
Formel: C₁₀H₈N₂O₂
Schmelzpunkt: 267…269 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00202955
CAS-Nummer: 387350-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR