6-Bromo-5-chloroindoline-2,3-dione

Lieferant: Apollo Scientific
Achtung

OR52245-250MG
APOSOR52245-250MGEA 142 EUR
APOSOR52245-250MG
6-Bromo-5-chloroindoline-2,3-dione
6-Bromo-5-chloroindoline-2,3-dione
Formel: C₈H₃BrClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10566349
CAS-Nummer: 192799-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR