1-Cyclohexyl-2-ethylbenzene 97%

Lieferant: Apollo Scientific

OR52259-1G
APOSOR52259-1GEA 63.8 EUR
APOSOR52259-1G
1-Cyclohexyl-2-ethylbenzene 97%
1-Cyclohexyl-2-ethylbenzene
Formel: C₁₄H₂₀
Molecular Weight: 188,309 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29090724
CAS-Nummer: 4501-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR