tert-Butyl-2,4-dioxopiperidine-1-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 2,4-dioxopiperidine-1-carboxylate, Piperidine-2,4-dione, N-BOC protected

OR52261-1G
APOSOR52261-1GEA 14.7 EUR
APOSOR52261-1G
tert-Butyl-2,4-dioxopiperidine-1-carboxylate
tert-Butyl-2,4-dioxopiperidine-1-carboxylate
Formel: C₁₀H₁₅NO₄
Molecular Weight: 213.2304 g/mol
Schmelzpunkt: 111…112 °C
Lagertemperatur: Kühlschrank
MDL: MFCD10566071
CAS-Nummer: 845267-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR