tert-Butyl 2-(piperidin-4-yl)pyrrolidine-1-carboxylate
Lieferant: Apollo Scientific
OR52273-1G
APOSOR52273-1GEA
836
EUR
APOSOR52273-1G
tert-Butyl 2-(piperidin-4-yl)pyrrolidine-1-carboxylate
tert-Butyl 2-(piperidin-4-yl)pyrrolidine-1-carboxylate
Formel:
C₁₄H₂₆N₂O₂ Molecular Weight: 254,368 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09702201 CAS-Nummer: 929974-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...