Methyl 4,4-dimethyl-2-oxocyclopentanecarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-oxo-4,4-dimethylcyclopentanecarboxylate, 4,4-Dimethyl-2-oxocyclopentanecarboxylic acid methyl ester

OR52274-1G
APOSOR52274-1GEA 1630 EUR
APOSOR52274-1G
Methyl 4,4-dimethyl-2-oxocyclopentanecarboxylate
Methyl 4,4-dimethyl-2-oxocyclopentanecarboxylate
Formel: C₉H₁₄O₃
Molecular Weight: 170,206 g/mol
Siedepunkt: 55…60 °C (2 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD07776759
CAS-Nummer: 60585-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR