Bicyclo[3.2.1]octan-3-one 95+%

Lieferant: Apollo Scientific
Achtung

OR52275-250MG
APOSOR52275-250MGEA 510 EUR
APOSOR52275-250MG
Bicyclo[3.2.1]octan-3-one 95+%
Bicyclo[3.2.1]octan-3-one
Formel: C₈H₁₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD19300751
CAS-Nummer: 14252-05-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR