2,2,6,6-Tetramethyloxan-4-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2,6,6-Tetramethyl-2H-3,5,6-trihydropyran-4-one, 2,2,6,6-Tetramethyltetrahydro-4H-pyran-4-one

OR52276-5G
APOSOR52276-5GEA 865 EUR
APOSOR52276-5G
2,2,6,6-Tetramethyloxan-4-one
2,2,6,6-Tetramethyloxan-4-one
Formel: C₉H₁₆O₂
Molecular Weight: 156,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12828096
CAS-Nummer: 1197-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR