Methyl 3-hydroxyazetidine-1-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR52278-10G
OR52278-1G
OR52278-5G
APOSOR52278-10GEA
852
EUR
APOSOR52278-10G
APOSOR52278-1G
APOSOR52278-5G
Methyl 3-hydroxyazetidine-1-carboxylate
Methyl 3-hydroxyazetidine-1-carboxylate
Formel:
C₅H₉NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18072727 CAS-Nummer: 118972-97-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...