N-Benzhydryl-N-methylamine

Lieferant: Apollo Scientific
Gefahr

OR5228-500MG
APOSOR5228-500MGEA 21.9 EUR
APOSOR5228-500MG
N-Benzhydryl-N-methylamine
N-Benzhydryl-N-methylamine
Formel: C₁₄H₁₅N
Schmelzpunkt: 38…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00467853
CAS-Nummer: 14683-47-7
UN: 3259
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR