5-Chloropyridazin-3-(2H)-one 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloropyridazin-3(2H)-one

OR52286-1G
APOSOR52286-1GEA 78.3 EUR
APOSOR52286-1G
5-Chloropyridazin-3-(2H)-one 97%
5-Chloropyridazin-3-(2H)-one
Formel: C4H3ClN2O
Molecular Weight: 130,5300 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12198134
CAS-Nummer: 660425-07-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR