(S)-1-(1H-Indazol-5-yl)ethanamine

Lieferant: Apollo Scientific

OR52291-500MG
APOSOR52291-500MGEA 466 EUR
APOSOR52291-500MG
(S)-1-(1H-Indazol-5-yl)ethanamine
(S)-1-(1H-Indazol-5-yl)ethanamine
Formel: C₉H₁₁N₃
Molecular Weight: 161,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18209561
CAS-Nummer: 1259610-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR