(S)-1-(1H-Pyrazolo[3,4-b]pyridin-5-yl)ethanamine
Lieferant: Apollo Scientific
OR52293-500MG
APOSOR52293-500MGEA
466
EUR
APOSOR52293-500MG
(S)-1-(1H-Pyrazolo[3,4-b]pyridin-5-yl)ethanamine
(S)-1-(1H-Pyrazolo[3,4-b]pyridin-5-yl)ethanamine
Formel:
C₈H₁₀N₄ Molecular Weight: 162,192 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29042486 CAS-Nummer: 1932616-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...