(S)-1-(Isoquinolin-8-yl)ethanamine

Lieferant: Apollo Scientific

OR52296-500MG
APOSOR52296-500MGEA 466 EUR
APOSOR52296-500MG
(S)-1-(Isoquinolin-8-yl)ethanamine
(S)-1-(Isoquinolin-8-yl)ethanamine
Formel: C₁₁H₁₂N₂
Molecular Weight: 172,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21348027
CAS-Nummer: 1344594-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR