(S)-1-(Furo[2,3-b]pyridin-5-yl)ethanamine

Lieferant: Apollo Scientific

OR52298-500MG
APOSOR52298-500MGEA 466 EUR
APOSOR52298-500MG
(S)-1-(Furo[2,3-b]pyridin-5-yl)ethanamine
(S)-1-(Furo[2,3-b]pyridin-5-yl)ethanamine
Formel: C₉H₁₀N₂O
Molecular Weight: 162,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29042488
CAS-Nummer: 1932107-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR