(S)-1-(5-Methylpyrimidin-2-yl)ethanamine

Lieferant: Apollo Scientific

OR52299-500MG
APOSOR52299-500MGEA 466 EUR
APOSOR52299-500MG
(S)-1-(5-Methylpyrimidin-2-yl)ethanamine
(S)-1-(5-Methylpyrimidin-2-yl)ethanamine
Formel: C₇H₁₁N₃
Molecular Weight: 137,182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24249248
CAS-Nummer: 1568163-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR