(S)-1-(5-Bromopyrimidin-2-yl)ethanamine

Lieferant: Apollo Scientific

OR52300-500MG
APOSOR52300-500MGEA 0 EUR
APOSOR52300-500MG
(S)-1-(5-Bromopyrimidin-2-yl)ethanamine
(S)-1-(5-Bromopyrimidin-2-yl)ethanamine
Formel: C₆H₈BrN₃
Molecular Weight: 202,052 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29042489
CAS-Nummer: 1447447-20-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR