(S)-1-(2,6-Dichloropyridin-3-yl)ethanamine

Lieferant: Apollo Scientific

OR52301-500MG
APOSOR52301-500MGEA 466 EUR
APOSOR52301-500MG
(S)-1-(2,6-Dichloropyridin-3-yl)ethanamine
(S)-1-(2,6-Dichloropyridin-3-yl)ethanamine
Formel: C₇H₈Cl₂N₂
Molecular Weight: 191,058 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09830322
CAS-Nummer: 1213971-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR