(S) -1-(5-Bromo-3-chloropyridin-2-yl)ethanamine

Lieferant: Apollo Scientific

OR52302-500MG
APOSOR52302-500MGEA 466 EUR
APOSOR52302-500MG
(S) -1-(5-Bromo-3-chloropyridin-2-yl)ethanamine
(S) -1-(5-Bromo-3-chloropyridin-2-yl)ethanamine
Formel: C₇H₈BrClN₂
Molecular Weight: 235,509 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18209623
CAS-Nummer: 793695-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR