(S)-1-(3-Bromo-5-chloropyridin-2-yl)ethanamine
Lieferant: Apollo Scientific
OR52304-500MG
APOSOR52304-500MGEA
466
EUR
APOSOR52304-500MG
(S)-1-(3-Bromo-5-chloropyridin-2-yl)ethanamine
(S)-1-(3-Bromo-5-chloropyridin-2-yl)ethanamine
Formel:
C₇H₈BrClN₂ Molecular Weight: 235,509 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18209621 CAS-Nummer: 1259882-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...