(S)-1-(5-Bromo-4-methylpyridin-2-yl)ethanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR52306-500MG
APOSOR52306-500MGEA 618 EUR
APOSOR52306-500MG
(S)-1-(5-Bromo-4-methylpyridin-2-yl)ethanamine hydrochloride
(S)-1-(5-Bromo-4-methylpyridin-2-yl)ethanamine hydrochloride
Formel: C₈H₁₁BrN₂·ClH
Molecular Weight: 251,551 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27924161
CAS-Nummer: 1628810-35-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR