tert-Butyl 4-hydroxy-1,4'-bipiperidine-1'-carboxylate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-hydroxy-[1,4'-bipiperidine]-1'-carboxylate, tert-Butyl 4-(4-hydroxypiperidin-1-yl)piperidine-1-carboxylate
OR52308-1G
APOSOR52308-1GEA
159
EUR
APOSOR52308-1G
tert-Butyl 4-hydroxy-1,4'-bipiperidine-1'-carboxylate 98%
tert-Butyl 4-hydroxy-1,4'-bipiperidine-1'-carboxylate
Formel:
C₁₅H₂₈N₂O₃ Molecular Weight: 284,394 g/mol Schmelzpunkt: 98…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12546285 CAS-Nummer: 367500-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...