5-Nitro-2-furaldehyd 98%
Lieferant: Apollo Scientific
Synonyme:
5-Nitrofurfural
OR5231-25G
APOSOR5231-25GEA
65.4
EUR
APOSOR5231-25G
5-Nitro-2-furaldehyd 98%
5-Nitro-2-furaldehyd
Formel:
C₅H₃NO₄ Molecular Weight: 141,08 g/mol Siedepunkt: 121 °C/10mmHg Schmelzpunkt: 37…39 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003230 CAS-Nummer: 698-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...