3,3-Dimethyloxan-4-amine hydrochloride

Lieferant: Apollo Scientific

OR52315-1G
APOSOR52315-1GEA 611 EUR
APOSOR52315-1G
3,3-Dimethyloxan-4-amine hydrochloride
3,3-Dimethyloxan-4-amine hydrochloride
Formel: C₇H₁₅NO·ClH
Molecular Weight: 165,661 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28155170
CAS-Nummer: 1400580-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR