2-(4-Brombutyl)-1,3-dioxolan
Lieferant: Apollo Scientific
|
Achtung
|
OR52318-1G
APOSOR52318-1GEA
144
EUR
APOSOR52318-1G
2-(4-Brombutyl)-1,3-dioxolan
2-(4-Bromobutyl)-1,3-dioxolane
Formel:
C₇H₁₃BrO₂ Dichte: 1,38 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093454 CAS-Nummer: 87227-41-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...