4,5,6,7-Tetrahydro[1,3]thiazolo[5,4-c]pyridine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,5,6,7-Tetrahydrothiazolo[5,4-c]pyridine hydrochloride

OR52323-250MG
APOSOR52323-250MGEA 437 EUR
APOSOR52323-250MG
4,5,6,7-Tetrahydro[1,3]thiazolo[5,4-c]pyridine hydrochloride
4,5,6,7-Tetrahydro[1,3]thiazolo[5,4-c]pyridine hydrochloride
Formel: C₆H₈N₂S·ClH
Molecular Weight: 176,667 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08726044
CAS-Nummer: 1241725-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR