L-tert-Leucin-Methylamid

Lieferant: Apollo Scientific
Achtung

Synonyme: L-tert-Leucylaminomethane, (S)-2-Amino-N,3,3-trimethylbutanamide

OR52327-1G
APOSOR52327-1GEA 184 EUR
APOSOR52327-1G
L-tert-Leucin-Methylamid
L-tert-Leucin-Methylamid
Formel: C₇H₁₆N₂O
Molecular Weight: 144,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00673329
CAS-Nummer: 89226-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR