2-tert-Butyl-1H-imidazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(tert-Butyl)-1H-imidazole
OR52331-5G
APOSOR52331-5GEA
77
EUR
APOSOR52331-5G
2-tert-Butyl-1H-imidazole
2-tert-Butyl-1H-imidazole
Formel:
C₇H₁₂N₂ Molecular Weight: 124.19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16618480 CAS-Nummer: 36947-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...