3-Methoxy-2-methylbenzaldehyde 97%

Lieferant: Apollo Scientific

OR52335-1G
APOSOR52335-1GEA 91.5 EUR
APOSOR52335-1G
3-Methoxy-2-methylbenzaldehyde 97%
3-Methoxy-2-methylbenzaldehyde
Formel: C₉H₁₀O₂
Molecular Weight: 150,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044581
CAS-Nummer: 56724-03-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR