2-(Prop-1-en-2-yl)aniline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Isopropenylaniline
OR52354-5G
APOSOR52354-5GEA
92.8
EUR
APOSOR52354-5G
2-(Prop-1-en-2-yl)aniline
2-(Prop-1-en-2-yl)aniline
Formel:
C₉H₁₁N Molecular Weight: 133,19 g/mol Siedepunkt: 95 °C Flammpunkt: 78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007719 CAS-Nummer: 52562-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...