Cyclopropanecarboxylic acid anhydride

Lieferant: Apollo Scientific
Achtung

OR52359-10G OR52359-1G OR52359-5G
APOSOR52359-10GEA 742 EUR
APOSOR52359-10G APOSOR52359-1G APOSOR52359-5G
Cyclopropanecarboxylic acid anhydride
Cyclopropanecarboxylic acid anhydride
Formel: C₈H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12404941
CAS-Nummer: 33993-24-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR