2-Azaspiro[3.3]heptan-6-ol hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR52371-1G
APOSOR52371-1GEA 221 EUR
APOSOR52371-1G
2-Azaspiro[3.3]heptan-6-ol hydrochloride 97%
2-Azaspiro[3.3]heptan-6-ol hydrochloride
Formel: C₆H₁₁NO·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD23703064
CAS-Nummer: 1630907-10-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR