6-Chloro-4-pyrimidinecarboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloropyrimidine-4-carboxylic acid

OR52384-1G
APOSOR52384-1GEA 115 EUR
APOSOR52384-1G
6-Chloro-4-pyrimidinecarboxylic acid 97%
6-Chloro-4-pyrimidinecarboxylic acid
Formel: C₅H₃ClN₂O₂
Molecular Weight: 158,54 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08692928
CAS-Nummer: 37131-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR