3-(N-tert-Butoxycarbonyl-N-methylamino)aniline 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(N-Tert-butoxycarbonyl-N-methylamino)aniline, N-Methylbenzene-1,3-diamine, N-BOC protected

OR52385-1G
APOSOR52385-1GEA 154 EUR
APOSOR52385-1G
3-(N-tert-Butoxycarbonyl-N-methylamino)aniline 97%
3-(N-tert-Butoxycarbonyl-N-methylamino)aniline
Formel: C12H18N2O2
Molecular Weight: 222,28 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13677178
CAS-Nummer: 528882-16-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR