Aminoguanidin Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Aminoguanidine HCl, Aminoguanidine Hydrochloride, Guanylhydrazine hydrochloride
OR52394-100G
OR52394-500G
APOSOR52394-100GEA
123
EUR
APOSOR52394-100G
APOSOR52394-500G
Aminoguanidin Hydrochlorid
Aminoguanidin Hydrochlorid
Formel:
CH₇ClN₄ Molecular Weight: 110.55 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039074 CAS-Nummer: 1937-19-5 EINECS: 217-707-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...