1,4-Dichloropyrido[3,4-d]pyridazine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR52405-250MG
APOSOR52405-250MGEA
79.9
EUR
APOSOR52405-250MG
1,4-Dichloropyrido[3,4-d]pyridazine 97%
1,4-Dichloropyrido[3,4-d]pyridazine
Formel:
C₇H₃Cl₂N₃ Molecular Weight: 200.0300 g/mol Schmelzpunkt: 159 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09839782 CAS-Nummer: 14490-19-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...