Piperidino[2]piperidylmethane
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(Piperidin-2-ylmethyl)piperidine
OR52414-1G
APOSOR52414-1GEA
109
EUR
APOSOR52414-1G
Piperidino[2]piperidylmethane
Piperidino[2]piperidylmethane
Formel:
C₁₁H₂₂N₂ Molecular Weight: 182,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085502 CAS-Nummer: 81310-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...