1,3-Di-tert-butylimidazol-2-ylidene 98%

Lieferant: Apollo Scientific
Achtung

OR52422-1G
APOSOR52422-1GEA 226 EUR
APOSOR52422-1G
1,3-Di-tert-butylimidazol-2-ylidene 98%
1,3-Di-tert-butylimidazol-2-ylidene
Formel: C₁₁H₂₀N₂
Lagertemperatur: Kühlschrank
MDL: MFCD03788921
CAS-Nummer: 157197-53-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR