2-Bromo-5-methoxybenzyl bromide

Lieferant: Apollo Scientific
Gefahr

OR52426-100G OR52426-25G OR52426-5G
APOSOR52426-100GEA 190 EUR
APOSOR52426-100G APOSOR52426-25G APOSOR52426-5G
2-Bromo-5-methoxybenzyl bromide
2-Bromo-5-methoxybenzyl bromide
Formel: C₈H₈Br₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00078664
CAS-Nummer: 19614-12-1
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR