Fenuron
Lieferant: Apollo Scientific
|
Achtung
|
OR52463-1G
APOSOR52463-1GEA
161
EUR
APOSOR52463-1G
Fenuron
Fenuron
Formel:
C₉H₁₂N₂O Schmelzpunkt: 128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025662 CAS-Nummer: 101-42-8 EINECS: 202-941-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...