6-Chloro-2-methoxynicotinaldehyde 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-2-methoxypyridine-3-carbaldehyde

OR52468-5G
APOSOR52468-5GEA 119 EUR
APOSOR52468-5G
6-Chloro-2-methoxynicotinaldehyde 98%
6-Chloro-2-methoxynicotinaldehyde
Formel: C₇H₆ClNO₂
Molecular Weight: 171.581 g/mol
Schmelzpunkt: 78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11847276
CAS-Nummer: 95652-81-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR