N,N-Dimethyl-1,4-phenylendiammoniumdichlorid 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: N,N-Dimethyl-p-phenylenediamine dihydrochloride, N,N-Dimethyl-P-phenylenediamine 2HCl

OR5247-100G
APOSOR5247-100GEA 234 EUR
APOSOR5247-100G
N,N-Dimethyl-1,4-phenylendiammoniumdichlorid 95%
N,N-Dimethyl-1,4-phenylendiammoniumdichlorid
Formel: (CH₃)₂NC₆H₄NH₂·2HCl
Molecular Weight: 209,12 g/mol
Schmelzpunkt: 222 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00012991
CAS-Nummer: 536-46-9
EINECS: 208-635-7
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR