8-Methoxy-3,4-dihydro-2H-1-benzopyran-3-amine

Lieferant: Apollo Scientific
Achtung

OR52470-1G
APOSOR52470-1GEA 662 EUR
APOSOR52470-1G
8-Methoxy-3,4-dihydro-2H-1-benzopyran-3-amine
8-Methoxy-3,4-dihydro-2H-1-benzopyran-3-amine
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Schmelzpunkt: 37…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08544307
CAS-Nummer: 112904-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR