2-Naphthalinmethanol
Lieferant: Apollo Scientific
|
Achtung
|
OR52473-100G
OR52473-25G
OR52473-10G
APOSOR52473-100GEA
140
EUR
APOSOR52473-100G
APOSOR52473-25G
APOSOR52473-10G
2-Naphthalinmethanol
2-Naphthalinmethanol
Formel:
C₁₁H₁₀O Molecular Weight: 158.2000 g/mol Siedepunkt: 178 °C/7.5mmHg Schmelzpunkt: 79…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004124 CAS-Nummer: 1592-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...