1,2-Bis(2,4,6-tribromophenoxy)ethane 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,1-(1,2-Ethanediylbis(oxy))bis(2,4,6-tribromobenzene)
OR52482-1G
APOSOR52482-1GEA
78.3
EUR
APOSOR52482-1G
1,2-Bis(2,4,6-tribromophenoxy)ethane 97%
1,2-Bis(2,4,6-tribromophenoxy)ethane
Formel:
C₁₄H₈Br₆O₂ Molecular Weight: 687,636 g/mol Schmelzpunkt: 223,5…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00071811 CAS-Nummer: 37853-59-1 EINECS: 253-692-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...